Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-sulfo-link-biotin | 1363444-70-5 | >95.0% | 653.77 g/mol | C31H35N5O7S2 | 5 MG
SDP

Catalog No. 5000202733
Encompass_Preferred

DBCO-sulfo-link-biotin is a cleavable linker reagent that combines a dibenzocyclooctyne (DBCO) reactive group with a sulfonated biotin tag for aqueous solubility and affinity capture. It is intended for click chemistry applications such as strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-bearing biomolecules and for use in the synthesis of antibody-drug conjugates. For research use only.

  • Enables SPAAC with azide-functionalized biomolecules.
  • Contains a sulfonated biotin tag for aqueous solubility and affinity capture.
  • Cleavable linker suitable for antibody-drug conjugate synthesis.
  • Soluble in DMSO at high concentrations and formulatable for in vivo use.
  • Powder stable at -20°C; solutions stable at -80°C for extended storage.
  • Available in small milligram quantities for research workflows.

Catalog No. 50-002-02733 Supplier Medchemexpress LLC Supplier No. HY1308105MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-sulfo-link-biotin is a cleavable linker reagent that combines a dibenzocyclooctyne (DBCO) reactive group with a sulfonated biotin tag for aqueous solubility and affinity capture. It is intended for click chemistry applications such as strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-bearing biomolecules and for use in the synthesis of antibody-drug conjugates. For research use only.

  • Enables SPAAC with azide-functionalized biomolecules.
  • Contains a sulfonated biotin tag for aqueous solubility and affinity capture.
  • Cleavable linker suitable for antibody-drug conjugate synthesis.
  • Soluble in DMSO at high concentrations and formulatable for in vivo use.
  • Powder stable at -20°C; solutions stable at -80°C for extended storage.
  • Available in small milligram quantities for research workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.