Learn More
Broadpharm DBCO-Sulfo-NHS ester, 1400191-52-7, MFCD22572669, 25mg
Catalog No. NC1622716
Encompass
Formula: C25H21N2NaO8S, Purity: 97%, MW: 532.5, DBCO-Sulfo-NHS ester is a water-soluble sulfocated reagent containing DBCO moiety. It can be used for simple and efficient labeling of antibodies, proteins and any other primary amine-bearing macromolecules with DBCO moiety in 100% aqueous buffers.
Catalog No. NC1622716 Supplier Broadpharm Supplier No. BP2228925MG
Formula: C25H21N2NaO8S, Purity: 97%, MW: 532.5, DBCO-Sulfo-NHS ester is a water-soluble sulfocated reagent containing DBCO moiety. It can be used for simple and efficient labeling of antibodies, proteins and any other primary amine-bearing macromolecules with DBCO moiety in 100% aqueous buffers.