Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBD-CO-Hz 98.0+%, TCI America™
SDP

Catalog No. A5571100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5571100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5571100MG Supplier TCI America Supplier No. A5571100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[for HPLC Labeling]

Chemical Identifiers

CAS 179951-63-4
Molecular Formula C11H16N6O4S
Molecular Weight (g/mol) 328.35
MDL Number MFCD01321128
InChI Key CCERXXCKPHMFBQ-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-(N-hydrazinocarbonylmethyl-N-methyl)aminobenzofurazan, 4-(N,N-Dimethylaminosulfonyl)-7-(N-hydrazinocarbonylmethyl-N-methyl)amino-2,1,3-benzoxadiazole
PubChem CID 44630077
IUPAC Name 7-{[(hydrazinecarbonyl)methyl](methyl)amino}-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(N(C)CC(=O)NN)C2=NON=C12

Specifications

Melting Point 166°C
Quantity 100 mg
Formula Weight 328.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material DBD-CO-Hz [=4-(N,N-Dimethylaminosulfonyl)-7-(N-hydrazinocarbonylmethyl-N-methyl)amino-2,1,3-b
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.