Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBD-ED [=4-(N,N-Dimethylaminosulfonyl)-7-(2-aminoethylamino)-2,1,3-benzoxadiazole], TCI America™
SDP

Catalog No. A5574100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5574100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5574100MG Supplier TCI America Supplier No. A5574100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[for HPLC Labeling]

Chemical Identifiers

CAS 189373-41-9
Molecular Formula C10H15N5O3S
Molecular Weight (g/mol) 285.322
MDL Number MFCD01321129
InChI Key RDMBHRPODMXKHT-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-(2-aminoethylamino)-2,1,3-benzoxadiazole
PubChem CID 4125780
IUPAC Name 7-(2-aminoethylamino)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)NCCN

Specifications

Quantity 100 mg
Formula Weight 285.32
Physical Form Crystalline Powder
Chemical Name or Material DBD-ED [=4-(N,N-Dimethylaminosulfonyl)-7-(2-aminoethylamino)-2,1,3-benzoxadiazole] [for HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.