Learn More
Apexbio Technology LLC DBeQ, 177355-84-9, MFCD03691820, 10 mg
SDP
Catalog No. 501012372
Encompass_Preferred
MF: C22H20N4, MW: 340.42, Solubility: 16mg/mL in DMSO. DBeQ is a selective and reversible inhibitor of p97 ATPase (IC50 = 1.5 uM) which induces caspase-3 and caspase-7 rapidly.
Catalog No. 50-101-2372 Supplier Apexbio Technology LLC Supplier No. A862910
MF: C22H20N4, MW: 340.42, Solubility: 16mg/mL in DMSO. DBeQ is a selective and reversible inhibitor of p97 ATPase (IC50 = 1.5 uM) which induces caspase-3 and caspase-7 rapidly.