Learn More
TARGETMOL CHEMICALS INC DBET1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T449510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. dBET1 is a hybrid molecule that combines (+)-JQ1 and thalidomide. It induces cereblon-dependent BET protein degradation in vitro (EC50 430 nM) and induces apoptosis. purity: 99%
Catalog No. 50-257-9876