Learn More
Enzo Life Sciences dBET1 (10mg). CAS: 1799711-21-9
SDP
Catalog No. 502013719
Encompass_Preferred
dBET1 is a PROTAC, consisting of a BRD4-ligand JQ-1 linked to a cereblon E3 ligase ligand, targeting on BET bromodomain BRD4. Purity: ≥99%. Long Term Storage: -20°C.
Catalog No. 50-201-3719 Supplier Enzo Life Sciences Supplier No. ENZCHM2600010
dBET1 is a PROTAC, consisting of a BRD4-ligand JQ-1 linked to a cereblon E3 ligase ligand, targeting on BET bromodomain BRD4. Purity: ≥99%. Long Term Storage: -20°C.