Learn More
Medchemexpress LLC dBET1 | 1799711-21-9 | MFCD31544503 | 99.0% | 785.27 | C38H37ClN8O7S | 50 MG
SDP
Catalog No. 5000248598
Encompass_Preferred
dBET1 is a PROTAC research compound that induces targeted degradation of BRD4 by recruiting the Cereblon E3 ubiquitin ligase. It is used in studies of BET bromodomain biology and targeted protein degradation in preclinical research.
  • Induces Cereblon-mediated BRD4 degradation (EC50 ~430 nM).
  • Designed as a bifunctional molecule linking a bromodomain ligand to a Cereblon ligand.
  • High purity suitable for biochemical and cellular assays (≈99%).
  • Available as a white to light yellow powder for easy handling.
  • Stable when stored as a powder at -20°C for up to 3 years.
  • Packaged in small research quantities for assay development (50 mg available).
Catalog No. 50-002-48598 Supplier Medchemexpress LLC Supplier No. HY10183850MG
dBET1 is a PROTAC research compound that induces targeted degradation of BRD4 by recruiting the Cereblon E3 ubiquitin ligase. It is used in studies of BET bromodomain biology and targeted protein degradation in preclinical research.
  • Induces Cereblon-mediated BRD4 degradation (EC50 ~430 nM).
  • Designed as a bifunctional molecule linking a bromodomain ligand to a Cereblon ligand.
  • High purity suitable for biochemical and cellular assays (≈99%).
  • Available as a white to light yellow powder for easy handling.
  • Stable when stored as a powder at -20°C for up to 3 years.
  • Packaged in small research quantities for assay development (50 mg available).