Learn More
TARGETMOL CHEMICALS INC DBET57 5MG
SDP
Catalog No. 502576682
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. dBET57 is heterobifunctional degrader of BRD4 based on the PROTAC technology exhibits significant degradation of BRD4BD1 with a DC50/5h of 500 nM and inactive on BRD4BD2purity: 99%
Catalog No. 50-257-6682 Supplier TARGETMOL CHEMICALS INC Supplier No. T54405MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. dBET57 is heterobifunctional degrader of BRD4 based on the PROTAC technology exhibits significant degradation of BRD4BD1 with a DC50/5h of 500 nM and inactive on BRD4BD2purity: 99%