Learn More
Medchemexpress LLC HY-123844 5mg Medchemexpress, dBET57 CAS:1883863-52-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1238445MG
Encompass_Preferred
Medchemexpress, HY-123844 5mg dBET57 CAS:1883863-52-2 dBET57 is a potent and selective degrader of BRD4 BD1 based on the PROTAC technology. dBET57 mediates recruitment to the CRL4 CRBN E3 ubiquitin ligase, with a DC 50/5h of 500 nM for BRD4 BD1 , and is inactive on BRD4 BD2 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2583