Learn More
Medchemexpress LLC HY-112588 10mg , dBET6 CAS:1950634-92-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11258810MG
Encompass_Preferred
HY-112588 10mg dBET6 CAS: 1950634-92-0 dBET6 is a highly potent, selective and cell-permeable PROTAC connected by ligands for Cereblon and BET, with an IC50 of 14 nM, and has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9093