Learn More
TARGETMOL CHEMICALS INC DBET6 10MG
SDP
Catalog No. 502579806
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. dBET6 is a selective and cell-permeable degrader of BET based on PROTAC (IC50 14 nM). It has antitumor activity. purity: 99%
Catalog No. 50-257-9806 Supplier TARGETMOL CHEMICALS INC Supplier No. T513010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. dBET6 is a selective and cell-permeable degrader of BET based on PROTAC (IC50 14 nM). It has antitumor activity. purity: 99%