Learn More
Enzo Life Sciences dBET6 (50mg). CAS: 1950634-92-0
SDP
Supplier:  Enzo Life Sciences ENZCHM2570050
Encompass_Preferred
dBET6 is chemical degrader of BET bromodomain proteins based on PROTAC and led a collapse of global elongation that phenocopies CDK9 inhibition. Purity: ≥99%. Solubility: Soluble in DMSO (240 mg/mL). Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-3710