Learn More
Selleck Chemical LLC dBET6 5mg 1950634-92-0
SDP
Catalog No. 501948252
Encompass_Preferred
dBET6 is a highly cell-permeable PROTAC degrader of BET bromodomains with an IC50 of 14 nM for BRD4 binding. dBET6 also induces c-MYC downregulation and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8252 Supplier Selleck Chemical LLC Supplier No. S87625MG
dBET6 is a highly cell-permeable PROTAC degrader of BET bromodomains with an IC50 of 14 nM for BRD4 binding. dBET6 also induces c-MYC downregulation and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.