Learn More
Aviva Systems Biology Corp DBI Antibody - N-terminal region (ARP33135_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33135_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DBI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DBI. Alternate names: ACBD1, ACBP, CCK-RP, EP, MGC70414ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54894