Learn More
MSC DBI-SALA 3M Adjustable Offset Davit Mast, Confined Space, 1 ea
Catalog No. 5000781446
Encompass
The 3M™ DBI-SALA® Confined Space Adjustable Offset Upper Davit Mast is a durable, high-capacity solution for rescue and hoisting workers. This versatile mast is designed for strength and efficiency, ideal for confined spaces, and can be customized with up to two Lower Mast Extensions to adjust the overall height of your system. It is fall arrest rated in all davit offset mast positions, ensuring safety and accessibility, and features a lightweight, portable design for ease of use.
  • Integrated 5,000 lb. (22 kN) anchorage point
  • Fall arrest rated in all davit offset mast positions
  • Combines with lower mast extensions for height adjustment
  • Two extension ranges: 11.5 in. to 27.5 in. and 23.5 in. to 42.5 in.
  • Integrated secondary fall arrest anchorage
  • For use with portable and permanent bases
  • Built-in primary and secondary pulleys
Catalog No. 50-007-81446 Supplier MSC Supplier No. 53869137
The 3M™ DBI-SALA® Confined Space Adjustable Offset Upper Davit Mast is a durable, high-capacity solution for rescue and hoisting workers. This versatile mast is designed for strength and efficiency, ideal for confined spaces, and can be customized with up to two Lower Mast Extensions to adjust the overall height of your system. It is fall arrest rated in all davit offset mast positions, ensuring safety and accessibility, and features a lightweight, portable design for ease of use.
  • Integrated 5,000 lb. (22 kN) anchorage point
  • Fall arrest rated in all davit offset mast positions
  • Combines with lower mast extensions for height adjustment
  • Two extension ranges: 11.5 in. to 27.5 in. and 23.5 in. to 42.5 in.
  • Integrated secondary fall arrest anchorage
  • For use with portable and permanent bases
  • Built-in primary and secondary pulleys