Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ 5 lb. Tool Retractor

Catalog No. 19809508 Shop All 3M Products
Change view
Click to view available options
Material:
Zytel, Aluminum, Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-809-508 Zytel, Aluminum, Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-809-508 Supplier 3M™ Supplier No. 7100214237
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ DBI-SALA™ 5 lb. Tool Retractor designed to minimize tangling and tripping hazards

  • Retractable design minimizes tangling and tripping hazards
  • 2:1 Safety Factor
  • Third party certified to a 5 lb. (2.3 kg) maximum capacity
  • 10.25 - 50 in. working range (26.04 - 127 cm)
  • Self-locking aluminum carabiners
  • Lightweight and compact design
  • Swiveling top housing connection
  • Reduces weight on workers' belts and harnesses
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Material Zytel, Aluminum, Steel
Includes 1/Pk.
Weight (English) 0.36 lb.
Load Bearing Capacity (Metric) 2.3 kg
Product Type Tool Retractor
Length (Metric) 19.05 cm
Weight (Metric) 0.18 kg
Length (English) 7.5 in.
Load Bearing Capacity (English) 5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.