Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Adjustable Radio/Cell Phone Holster with Clip2Loop Coil and Micro D-Ring

Catalog No. 19070796 Shop All 3M Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-070-796 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-070-796 Supplier 3M™ Supplier No. 7100157155
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M DBI-SALA™ Adjustable Radio/Cell Phone Holster with Clip2Loop Coil adjusts to the size of virtually any portable radio or cell phone

  • D-Ring connection point
  • Can be used from a belt or harness
  • Ergonomic quick-cinch ergonomic cord lock
  • Includes Clip2Loop Coil Tether and Micro D-Ring
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Width (English) 2.25 in.
For Use With (Equipment) Tools
Product Type Adjustable holster
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.