Learn More
3M™ DBI-SALA™ Advanced™ 5-Piece Davit Hoist System
Catalog No. 19070826 Shop All 3M Products
Change view
Click to view available options
Includes:
11-1/2 in. to 27-1/2 in. (29.2 to 69.8 cm) adjustable offset upper mast, 33 in. (83.8 cm) lower davit mast extension, 3-piece lightweight portable base, and winch mount bracket
23-1/2 in. to 42-1/2 in. (59.7 to 108 cm) adjustable offset upper mast, 33 in. (83.8 cm) lower davit mast extension, 3-piece portable base, and winch mount bracket
Weight (English):
83.2 lb.
164.7 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Weight (English)
19-070-826 11-1/2 in. to 27-1/2 in. (29.2 to 69.8 cm) adjustable offset upper mast, 33 in. (83.8 cm) lower davit mast extension, 3-piece lightweight portable base, and winch mount bracket 83.2 lb.
19-070-828 23-1/2 in. to 42-1/2 in. (59.7 to 108 cm) adjustable offset upper mast, 33 in. (83.8 cm) lower davit mast extension, 3-piece portable base, and winch mount bracket 164.7 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-826 Supplier 3M™ Supplier No. 7100188840
Add to Cart
Add to Cart
Request a Quote
Designed for manhole and confined space entry/retrieval applications. The davit pivots for ease-of-rescue and has adjustment for overhead clearance restrictions.
  • Includes everything needed to build your confined space rescue and work support system
  • Adjustable length of davit ensures mechanical device lifeline(s) are always centered overdo the confined space
  • Route lifelines of two mechanical devices for smooth, efficient rescue and work support operations
  • Allows user to adjust system quickly and easily to site configuration without tools and collapse for transport and storage
  • Attach 2 extensions together in any combination to customise your davit system to fit your changing work area for ultimate flexibility
  • Set up and break down quickly
  • Unique foot level and height adjustment system
  • An ideal confined space support structure for one or more mechanical devices
  • One system to cover a range of different confined space scenarios
  • Extremely fast and safe mechanical device installation
  • Flexibility to change overall mast height and overall anchorage height
  • Unique adjustable base
  • Lightweight, corrosion resistant aluminum construction
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Certifications/Compliance ANSI Z359.1, OSHA 1926.502
Product Type Portable Install
Includes 11-1/2 in. to 27-1/2 in. (29.2 to 69.8 cm) adjustable offset upper mast, 33 in. (83.8 cm) lower davit mast extension, 3-piece lightweight portable base, and winch mount bracket
Weight (English) 83.2 lb.
Load Bearing Capacity (Metric) 204 kg
Weight (Metric) 37.70 kg