Learn More
MSC DBI-SALA Cable Horizontal Lifeline System, Galvanized, Temporary, 1 ea
Catalog No. 5000782681
Encompass
The Sayfline Cable Horizontal Lifeline System is an engineered fall protection solution designed for construction and general industrial applications. This all-metal system provides horizontal mobility and fall protection, is durable yet lightweight for easy transport, and is suitable for extended use in various environmental conditions.
  • Galvanized cable lifeline available in multiple lengths
  • Metal energy absorbers limit lifeline tension forces
  • Built-in, turnbuckle tensioner for system tensioning
  • All-metal design withstands environmental conditions
  • Accommodates two 310 lb (141 kg) workers per span
  • Equipped with RFID Wedge Grip termination device for quicker setup
  • Easy installation and horizontal design for free movement
  • Meets OSHA 1910.140 regulations
Catalog No. 50-007-82681 Supplier MSC Supplier No. 69567477
The Sayfline Cable Horizontal Lifeline System is an engineered fall protection solution designed for construction and general industrial applications. This all-metal system provides horizontal mobility and fall protection, is durable yet lightweight for easy transport, and is suitable for extended use in various environmental conditions.
  • Galvanized cable lifeline available in multiple lengths
  • Metal energy absorbers limit lifeline tension forces
  • Built-in, turnbuckle tensioner for system tensioning
  • All-metal design withstands environmental conditions
  • Accommodates two 310 lb (141 kg) workers per span
  • Equipped with RFID Wedge Grip termination device for quicker setup
  • Easy installation and horizontal design for free movement
  • Meets OSHA 1910.140 regulations