Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Canvas Parts Pouch, Small

Catalog No. 19070798 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Tethering and securing tools
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
19-070-798 Tethering and securing tools
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-070-798 Supplier 3M™ Supplier No. 7100156938
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M DBI-SALA™ Canvas Parts Pouch, Small is an innovative self-closure system which can be used from a belt or harness

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Duck Canvas
Product Type Small Parts Pouch
For Use With (Application) Tethering and securing tools
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.