Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Clip2Loop Coil Tether

Catalog No. 19070803 Shop All 3M Products
Change view
Click to view available options
DBI-SALA™ Clip2Loop Coil Tether
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-070-803 DBI-SALA™ Clip2Loop Coil Tether
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-803 Supplier 3M™ Supplier No. 7100239799/DEL

missing translation for 'validMainframeMsg'

3M DBI-SALA™ Clip2Loop Coil Tether Tool is an ultra compact coil style tether with a clip at one end and a loop at the other

TRUSTED_SUSTAINABILITY

Specifications

Product Type Clip2Loop Coil Tether
Load Bearing Capacity (English) 2 lb.
For Use With (Equipment) Tools
Load Bearing Capacity (Metric) 0.9 kg
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.