This confined space winch is designed to maximize efficiency and safety in challenging fall protection and confined space situations. It features a triple-redundant braking system, including three independent working pawls and a backup brake for enhanced security, serving as the primary means of support for a hoist system.
- Triple redundant braking system and brake wear indicator
- Features 9:1 and 4:1 gear ratio mechanical advantage
- Incorporated mounting bracket for tripods and davits
- Built-in digital usage indicator for service intervals
- Swiveling snap hook with impact indicator
- Integral carrying handle for transport
- Quick-mount brackets for easy attachment and detachment
- Maximum working load of 450 lb (204 kg)
- Includes 88 m (290 ft) galvanized steel cable
- Cast aluminum housing with powder coat paint finish