Learn More
MSC DBI-SALA Davit Hoist System, 5-Piece, Aluminum, 1 ea
Catalog No. 5000796659
Encompass
The Davit Hoist System is engineered for safe manhole and confined space entry and retrieval. Constructed from durable, lightweight, high-strength aluminum, it features a pivoting davit for optimal rescue angles and simple adjustment for overhead clearance challenges.
  • Designed for manhole and confined space entry and retrieval
  • Durable, lightweight construction
  • Made with high-strength aluminum
  • Davit pivots for improved rescue angles
  • Adjusts for overhead clearance restrictions
  • Integrated 5,000 lb (22 kN) mast anchorage point
  • Fall arrest rated in all davit offset mast positions
  • Adjustable mast height and base width
  • Adjustable foot-leveling system
  • Available in two extension ranges: 11.5 to 27.5 in (29.2 to 69.8 cm), and 23.5 to 42.5 in (59.7 to 108 cm)
  • Built-in level indicator
  • Easy to set-up and dismantle without tools
Catalog No. 50-007-96659 Supplier MSC Supplier No. 18655886
The Davit Hoist System is engineered for safe manhole and confined space entry and retrieval. Constructed from durable, lightweight, high-strength aluminum, it features a pivoting davit for optimal rescue angles and simple adjustment for overhead clearance challenges.
  • Designed for manhole and confined space entry and retrieval
  • Durable, lightweight construction
  • Made with high-strength aluminum
  • Davit pivots for improved rescue angles
  • Adjusts for overhead clearance restrictions
  • Integrated 5,000 lb (22 kN) mast anchorage point
  • Fall arrest rated in all davit offset mast positions
  • Adjustable mast height and base width
  • Adjustable foot-leveling system
  • Available in two extension ranges: 11.5 to 27.5 in (29.2 to 69.8 cm), and 23.5 to 42.5 in (59.7 to 108 cm)
  • Built-in level indicator
  • Easy to set-up and dismantle without tools