Learn More
Description
- Extremely compact and lightweight
- Exclusive Hi-10™ Vectran™ energy absorption system
- Dual-layer high strength/abrasion resistant webbing
- User friendly self locking snap hooks at each end
- Built-in reflective materials
- Impact indicator
Specifications
Specifications
| Webbing Material | Polyester Web |
| Length (English) | 6 ft. |
| Load Bearing Capacity (English) | 130 to 310 lb. |
| Length (Metric) | 1.8 m |
| Leg Configuration | 6 ft. (1.8 m) single-leg lanyard |
| Certifications/Compliance | ANSI A10.32, OSHA 1926.502, ANSI Z359.1, ANSI Z359.13, OSHA 1910.66 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.