Learn More
MSC DBI-SALA Loop Rebar HLL Stanchion, 1 EA
Catalog No. 5000796162
Encompass
Engineered for Horizontal Lifeline Systems, this temporary cable Loop Rebar Stanchion with Base offers easy installation, reliable performance, and flexibility. Designed for use with HLL Rebar systems, it is tool-free to install and endures harsh environments.
  • Base adjusts from beam top
  • Portable, lightweight design
  • Equipped with RFID identification tag for tracking
  • Recommended for construction and general industrial use
  • For use with additional personal protective equipment
  • Single-span systems support up to 2 workers; multi-span up to 6
  • Functions as a temporary horizontal lifeline system
  • Metal Energy shock absorbers reduce fall force
  • Meets OSHA regulations
Catalog No. 50-007-96162 Supplier MSC Supplier No. 23206055
Engineered for Horizontal Lifeline Systems, this temporary cable Loop Rebar Stanchion with Base offers easy installation, reliable performance, and flexibility. Designed for use with HLL Rebar systems, it is tool-free to install and endures harsh environments.
  • Base adjusts from beam top
  • Portable, lightweight design
  • Equipped with RFID identification tag for tracking
  • Recommended for construction and general industrial use
  • For use with additional personal protective equipment
  • Single-span systems support up to 2 workers; multi-span up to 6
  • Functions as a temporary horizontal lifeline system
  • Metal Energy shock absorbers reduce fall force
  • Meets OSHA regulations