Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Medium Tape Measure Sleeve

Catalog No. 19070797 Shop All 3M Products
Change view
Click to view available options
DBI-SALA™ Medium Tape Measure Sleeve
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-070-797 DBI-SALA™ Medium Tape Measure Sleeve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-797 Supplier 3M™ Supplier No. 7010595540

missing translation for 'validMainframeMsg'

3M DBI-SALA™ Medium Tape Measure Sleeve is designed for use with a Tape Measure Holster and conforms to many tape measure sizes up to 25ft. (7.62m)

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Load Bearing Capacity (Metric) 0.9 kg
Product Type Tape Measure Sleeve
Load Bearing Capacity (English) 2 lb.
For Use With (Equipment) Tools
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.