Learn More
3M™ DBI-SALA™ Nano-Lok™ Personal Self Retracting Lifeline 3100526, Web Steel Snap Hook
Catalog No. 17018262 Shop All 3M Products
Change view
Click to view available options
Material:
Zinc-plated Steel
Catalog No. Material
17-018-262 Zinc-plated Steel
1 options
Catalog No. 17-018-262 Supplier 3M™ Supplier No. 7100313161
Add to Cart
Add to Cart
3M™ DBI-SALA™ Nano-Lok™ Self-Retracting Lifeline is lightweight with a compact design and industrial-strength durability. 3M™ DBI-SALA™ Nano-Lok™ Self-Retracting Lifeline offers versatility for many different industrial applications.
  • Self-Retracting Lifeline is ideal for direct connection to most harnesses.
  • Whether the application requires single or twin leg configurations, mounting to an overhead anchor, or connection directly to a harness, there are numerous models to choose from that suit almost any application.
  • Self-Retracting Lifeline locks quickly stopping a fall within inches - providing more protection at low heights.
TRUSTED_SUSTAINABILITY
Specifications
Material Zinc-plated Steel
Length (Metric) 1.83 m
Color Black, Brown
Length (English) 6 ft.
Product Type Self-retracting Lifeline (Web)