Learn More
3M™ DBI-SALA™ Nano-Lok™ Tie-Back Twin-Leg Quick Connect Self Retracting Lifeline
Catalog No. 19040605 Shop All 3M Products
Change view
Click to view available options
lifeline, Tie-back, retracting, Twin-leg, DBI-SALA
Catalog No.
19-040-605 lifeline, Tie-back, retracting, Twin-leg, DBI-SALA
1 options
This item is not returnable. View return policy
Catalog No. 19-040-605 Supplier 3M™ Supplier No. 7012289690
This item is not returnable. View return policy
Designed for ease-of-use and is ideal for direct connection to most harnesses.
  • Ideal as lanyard replacement
  • i-Safe™ equipped
  • Twin-leg tie-off design
  • Built-in connector for single-leg SRL direct attachment to harness
  • Tough and impact resistant thermoplastic housing
  • Quick activating speed sensing brake system
  • High capacity external energy absorber and impact indicator
  • Wide variety of configurations available
  • Extremely compact, lightweight design
  • Swiveling anchorage loops
  • Tie-back wear layer web design
  • Tie-back self-locking hook on each lifeline
  • 2.7m (9 ft.) 19mm (0.75 in.) Dyeema™ fiber and polyester web lifeline
TRUSTED_SUSTAINABILITY
Specifications
Material Dynema Polyester with Nylon Tubular Web
Load Bearing Capacity (Metric) 190 kg
Length (Metric) 2.7 m
Length (English) 9 ft.
Certifications/Compliance OSHA 1926.502
Load Bearing Capacity (English) 420 lb.