Learn More
3M™ DBI-SALA™ Pole Climber's Adjustable Rope Positioning Lanyard
Catalog No. 19070800 Shop All 3M Products
Change view
Click to view available options
Connector Types:
Aluminum Snap Hook and Carabiner
Zinc-plated Steel Snap Hook and Carabiner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
19-070-800 Aluminum Snap Hook and Carabiner
19-070-799 Zinc-plated Steel Snap Hook and Carabiner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-800 Supplier 3M™ Supplier No. 7100239628
  • $865.00
    $796.65 / Each of 1
    Save $68.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Designed without shock absorbers, making them ideal for positioning or restraint applications. The self-locking snap hooks are preferred by safety professionals and workers alike. Specifically designed to allow you to open and connect the hook without getting your thumb or fingers in the way.
  • 8 ft. (2.4 m) adjustable rope positioning lanyard
  • Ideal for work positioning while pole climbing
  • Durable kernmantle rope with wear indicator
  • Abrasion resistant sleeve
  • User friendly self locking snap hook at one end
  • Lightweight and durable rope adjuster with carabiner at one end
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Material Nylon, Polyester, Kernmantle
Webbing Material Nylon Core/Polyester Cover Kernmantle Rope
Includes Rope adjuster
Length (English) 8 ft.
Certifications/Compliance OSHA 1926.502, OSHA 1926.959, OSHA 1910.268 (g)
Load Bearing Capacity (English) 310 lb.
Connector Types Aluminum Snap Hook and Carabiner
Length (Metric) 2.4 m