Learn More
3M™ DBI-SALA™ Quick Wrap Tape II
Catalog No. 19070774 Shop All 3M Products
Change view
Click to view available options
DBI-SALA™ Quick Wrap Tape II
Catalog No.
19-070-774 DBI-SALA™ Quick Wrap Tape II
1 options
Catalog No. 19-070-774 Supplier 3M™ Supplier No. 7100204962
Add to Cart
Add to Cart
3M DBI-SALA™ Quick Wrap Tape II creates instant tethering points
  • Redesigned Quick-Wrap Tape II helps deliver long-lasting tool attachment performance
  • Resists tears better
  • Stretches further
  • Quick and easy installation
  • Stays out of the way
  • Reduces tool downtime
Certifications
Capital Safety Gen. Mfg. Req.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Tethering and securing tools
Width (English) 2.25 in.
Product Type Quick Wrap Tape
Length (English) 9 ft.
Certifications/Compliance Capital Safety Gen. Mfg. Req.