Learn More
3M™ DBI-SALA™ Rescue/Retrieval Y-Lanyard with Spreader Bar
Catalog No. 19809708 Shop All 3M Products
Change view
Click to view available options
Material:
Zinc-plated Steel, Polyester Web
Catalog No. Material
19-809-708 Zinc-plated Steel, Polyester Web
1 options
Catalog No. 19-809-708 Supplier 3M™ Supplier No. 7012691244
  • $360.00
    $331.65 / Case of 1
    Save $28.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
3M™ DBI-SALA™ Rescue/Retrieval Y-Lanyard with Spreader Bar features a D-ring at the center and snap hooks at leg ends.
  • Keeps victim upright during rescue
  • Abrasion resistant polyester webbing
  • Built-in spreader bar
  • User friendly self locking snap hooks
  • High strength D-ring tie-off point
  • Equipped with 3M™ Connected Safety ID (CSID)
TRUSTED_SUSTAINABILITY
Specifications
Material Zinc-plated Steel, Polyester Web
Color Blue
Certifications/Compliance OSHA 1910.66, ANSI Z359.3, OSHA 1926.502
Length (English) 4 ft.
Length (Metric) 1.2 m
Load Bearing Capacity (English) 310 lb.
Product Type Rescue/Retrieval Y-Lanyard with Spreader Bar
Load Bearing Capacity (Metric) 141 kg