Learn More
3M™ DBI-SALA™ Nylon Rope Lanyard
Catalog No. 19809275 Shop All 3M Products
Change view
Click to view available options
Length (English):
6 ft.
50 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-809-275 50 ft.
18-841 6 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-809-275 Supplier 3M™ Supplier No. 1232470
Nylon rope single-leg with snap hooks at each end.
  • Single-leg lanyard
  • Ideal for work positioning or restraint
  • 1/2 in. (1.3 cm) three-strand filament nylon rope
  • Exclusive five-tuck splice and seizing method
  • User friendly self locking snap hooks at each end
  • Equipped with i-Safe™
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Self-Locking Snap Hooks at Both Ends
Certifications/Compliance Meets or exceeds all applicable ANSI and OSHA standards.
Length (English) 50 ft.