This temporary cable horizontal lifeline system is designed for concrete rebar or shear studs, fitting diameters from 1/2 in. to 1 in. and 4 in. to 12 in. center to center. It includes two stanchions with tie-back and a lifeline assembly, featuring a modular design to accommodate various steel and concrete beams. Spans extend up to 60 ft., configurable for indefinite lengths with optional bypass brackets.
- Complete 50 ft. (15.2 m) system includes two stanchions and cable assemblies
- Adjustable stanchion can be offset 20 degrees or set horizontal for transport
- Rugged 3/8 in. (9.5 mm) galvanized cable lifeline
- Metal energy absorber limits fall arrest forces
- Fits 1/2 in. to 1 in. diameter rebar and 4 in. to 12 in. center to center
- Flexible capacity with single or multi-span configurations
- Equipped with RFID and top mounted adjustment system