Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ ShockWave™2 100% Tie-Off Shock Absorbing Lanyard

Catalog No. 19070730 Shop All 3M Products
Change view
Click to view available options
Material:
Aluminum
Zinc-plated Steel
Certifications/Compliance:
ANSI A10.32, OSHA 1926.502, OSHA 1910.66
n/a
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Certifications/Compliance
19-070-730 Zinc-plated Steel ANSI A10.32, OSHA 1926.502, OSHA 1910.66
19-070-729 Aluminum n/a
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-730 Supplier 3M™ Supplier No. 7100188780
  • $665.00
    $611.65 / Each of 1

    Save $53.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unique stretchable design offers complete freedom of movement while reducing trip-fall hazards and dragging or snagging by expanding to 6 ft. (1.8 m) when taut and automatically contracting to 4-1/2 ft. (1.4 m) in reaction to the worker’s movements.

  • 6 ft. (1.8 m) double-leg 100% tie-off lanyard
  • Stretches in reaction to your movements
  • Abrasion resistant polyester tubular webbing
  • User friendly self locking snap hook at center
  • Self-locking rebar hook at leg ends
  • Energy absorbing design limits fall arrest forces
  • Built-in impact indicator
  • Protected labels, equipped with i-Safe™
TRUSTED_SUSTAINABILITY

Specifications

Material Zinc-plated Steel
Color Yellow
Webbing Material Polyester Tubular Web
Load Bearing Capacity (Metric) 59 to 141 kg
Load Bearing Capacity (English) 130 to 310 lb.
Leg Configuration Double-Leg 100% Tie-Off
Product Type 100% Tie-Off Shock Absorbing Lanyard
Arresting Force (English) Energy Absorber Class: 6 ft (1.8 m) Free Fall/MAAF 900 lb. (4 kN)
Length (English) 6 ft.
Certifications/Compliance ANSI A10.32, OSHA 1926.502, OSHA 1910.66
Length (Metric) 1.8 m
Connector Types Rebar Hook
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.