Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ ShockWave™2 100% Tie-Off Shock Absorbing Lanyard - Rescue

Catalog No. 19070802 Shop All 3M Products
Change view
Click to view available options
Load Bearing Capacity (English):
130 to 310 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (English)
19-070-802 130 to 310 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-802 Supplier 3M™ Supplier No. 7100207452
  • $688.00
    $633.65 / Each of 1

    Save $54.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unique stretchable design offers complete freedom of movement while reducing trip-fall hazards and dragging or snagging by expanding to 6 ft. (1.8 m) when taut and automatically contracting to 4-1/2 ft. (1.4 m) in reaction to the worker’s movements.

  • 6 ft. (1.8 m) double-leg 100% tie-off lanyard
  • Stretches in reaction to your movements
  • Integrated D-ring for SRL attachment or rescue
  • Abrasion resistant polyester tubular webbing
  • User friendly self locking snap hook at center
  • Self-locking steel rebar hook at leg ends
  • Energy absorbing design limits fall arrest forces
  • Built-in impact indicator
  • Protected labels, equipped with i-Safe™
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Material Zinc-plated Steel
Webbing Material Polyester Tubular Web
Leg Configuration Double-Leg 100% Tie-Off
Includes double-leg 100% tie-off with elastic web, D-ring for SRL or rescue and snap hook at center, steel rebar hooks at leg ends
Product Type 100% Tie-Off Rescue Shock Absorbing Lanyard
Load Bearing Capacity (English) 130 to 310 lb.
Length (Metric) 1.8 m
Certifications/Compliance OSHA 1910.66, OSHA 1926.502, ANSI A10.32
Arresting Force (English) Energy Absorber Class: 6 ft (1.8 m) Free Fall/MAAF 900 lb. (4 kN)
Connector Types Rebar Hook and Snap Hook
Length (English) 6 ft.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.