Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ ShockWave™2 Arc Flash Rated 100% Tie-Off Shock Absorbing Lanyard

Catalog No. 19070757 Shop All 3M Products
Change view
Click to view available options
Material:
Zinc-plated Steel, Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-070-757 Zinc-plated Steel, Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-757 Supplier 3M™ Supplier No. 1244632

missing translation for 'validMainframeMsg'

Unique stretchable design offers complete freedom of movement while reducing trip-fall hazards and dragging or snagging by expanding to 6 ft. (1.8 m) when taut and automatically contracting to 4-1/2 ft. (1.4 m) in reaction to the worker’s movements.

  • Arc flash rated
  • 6 ft. (1.8 m) double-leg 100% tie-off lanyard
  • Stretches in reaction to your movements
  • Nomex™/Kevlar™ fiber webbing
  • User friendly self locking snap hook at one end
  • Self-locking aluminum rebar hooks at leg ends
  • Energy absorbing design limits fall arrest forces
  • Built-in impact indicator
  • Protected labels, equipped with i-Safe™
TRUSTED_SUSTAINABILITY

Specifications

Material Zinc-plated Steel, Aluminum
Color Black
Load Bearing Capacity (Metric) 59 to 141 kg
Webbing Material Nomex™, Dupont™ Kevlar™ aramid fiber
Includes double-leg 100% tie-off with elastic Nomex™/Dupont™ Kevlar™ aramid fiber fiber webbing and snap hooks at center, aluminum rebar hooks at leg ends
Load Bearing Capacity (English) 130 to 310 lb.
Leg Configuration Double-Leg 100% Tie-Off
Length (English) 6 ft.
Product Type Arc Flash 100% Tie-Off Shock Absorbing Lanyard
Length (Metric) 1.8 m
Connector Types Rebar Hook and Snap Hook
Arresting Force (English) Energy Absorber Class: 6 ft (1.8 m) Free Fall/MAAF 900 lb. (4 kN)
Certifications/Compliance ASTM F887, ASTM F887-2005, OSHA 1910.66, OSHA 1926.502
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.