Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-Sala™ ShockWave 2™ Shock-Absorbing Lanyard

Catalog No. 19809959 Shop All 3M Products
Change view
Click to view available options
Connector Types:
Rebar Hook
Snap Hook
Product Type:
6 ft. (1.8 m) single-leg with tubular web and snap hook at one end, steel rebar hook at other end
6 ft. (1.8 m) single-leg with tubular web and snap hooks at each end
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Product Type
19-809-959 Snap Hook 6 ft. (1.8 m) single-leg with tubular web and snap hooks at each end
19-070-756 Rebar Hook 6 ft. (1.8 m) single-leg with tubular web and snap hook at one end, steel rebar hook at other end
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-809-959 Supplier 3M™ Supplier No. 7100157035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unique stretchable design offers complete freedom of movement while reducing trip-fall hazards and dragging or snagging by expanding to 6 ft. (1.8 m) when taut and automatically contracting to 4-1/2 ft. (1.4 m) in reaction to the worker’s movements.

  • 6 ft. (1.8 m) single-leg lanyard
  • Stretches in reaction to your movements
  • Abrasion resistant polyester tubular webbing
  • User friendly self locking snap hooks at each end
  • Energy absorbing design limits fall arrest forces
  • Built-in impact indicator
  • Protected labels, equipped with i-Safe™

Order Info

Available in a variety of styles.

Compliance

Meets ANSI™ Z359.7-2011 requirements.

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Material Zinc-plated Steel
Leg Configuration Single-Leg
Webbing Material Polyester Tubular Web
Product Type 6 ft. (1.8 m) single-leg with tubular web and snap hooks at each end
Load Bearing Capacity (English) 130 to 310 lb.
Length (Metric) 1.8 m
Arresting Force (English) Energy Absorber Class: 6 ft (1.8 m) Free Fall/MAAF 900 lb. (4 kN)
Connector Types Snap Hook
Certifications/Compliance ANSI Z359.1, OSHA 1926.502, OSHA 1910.66, ANSI Z359.13, ANSI A10.32
Length (English) 6 ft.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.