Learn More
Cayman Chemical DBIhumnrecombInt 100ug
SDP
Catalog No. 502945808
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged DBI expressed in E. coli • Amino acids: 1-87 (full length) • MW: 14 kDa
Catalog No. 50-294-5808 Supplier Cayman Chemical Supplier No. 42156100 UG
Source: Recombinant human N-terminal His-tagged DBI expressed in E. coli • Amino acids: 1-87 (full length) • MW: 14 kDa