Learn More
World Precision Instrument Double Banana Plug with Solder Turret Terminals
SDP
Catalog No. 50822068
Encompass
Standard connectors for electrophysiology applications
This connector has dual male banana connectors on one side and dual female banana connectors on the other. It also has turrent terminals for soldering. It does not have a cable.
Catalog No. 50-822-068 Supplier World Precision Instrument Supplier No. 3670
Standard connectors for electrophysiology applications
This connector has dual male banana connectors on one side and dual female banana connectors on the other. It also has turrent terminals for soldering. It does not have a cable.