Learn More
Electron Microscopy Sciences Orange Stick Double Bevel Ends
SDP
Catalog No. 5029749
Encompass_Preferred
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 7"
Diameter: 5/32"
Catalog No. 50-297-49 Supplier Electron Microscopy Sciences Supplier No. 7230410
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring and mixing and general laboratory applications.
OAL: 7"
Diameter: 5/32"