Learn More
MSC DBL COATED TAPE
Catalog No. 5000793205
Encompass
This double-coated tape features a 4 mil polyethylene film on a PET polyester carrier with a 60 lb densified kraft DK paper liner. It provides excellent UV, temperature, and humidity resistance, along with strong adhesion to various surfaces including high and low surface energy materials and metals.
  • Excellent UV resistance
  • Good adhesion to a wide variety of materials (HSE, LSE, and metals)
  • Good temperature and humidity resistance
  • High-performance splicing in printing applications
  • Good on films and foils
Catalog No. 50-007-93205 Supplier MSC Supplier No. 89916027
This double-coated tape features a 4 mil polyethylene film on a PET polyester carrier with a 60 lb densified kraft DK paper liner. It provides excellent UV, temperature, and humidity resistance, along with strong adhesion to various surfaces including high and low surface energy materials and metals.
  • Excellent UV resistance
  • Good adhesion to a wide variety of materials (HSE, LSE, and metals)
  • Good temperature and humidity resistance
  • High-performance splicing in printing applications
  • Good on films and foils