Learn More
Electron Microscopy Sciences Double Cooling Chamber for 0.2 ml tubes
SDP
Catalog No. 5033428
Encompass_Preferred
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe. Fits two 96-well PCR trays or each can hold up to (12) 0.5 ml and (8) 1.5 ml tubes.
Catalog No. 50-334-28 Supplier Electron Microscopy Sciences Supplier No. 6195510
Anodized aluminum blocks chambers are ideal for ice pans, buckets, cold or hot plates. Excellent temperature conductors. Do not fit CoolSafe. Fits two 96-well PCR trays or each can hold up to (12) 0.5 ml and (8) 1.5 ml tubes.