Learn More
Cole-Parmer Cole-Parmer High-pH Double-Junction pH Electrode, 10-ft Cable
Catalog No. 50633576 Shop All Cole-Parmer Products
Encompass_Preferred
High-pH, double-junction electrodes are well suited for pH levels above 13 or high sodium ion concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Maximum pressure is 150 psi at 25°>C.
Catalog No. 50-633-576 Supplier Cole-Parmer Supplier No. 0599423
High-pH, double-junction electrodes are well suited for pH levels above 13 or high sodium ion concentrations. Double-junction reference protects against contamination from heavy metals, organics, and sulfides. Maximum pressure is 150 psi at 25°>C.