Learn More
MSC SEYMOUR-MIDWEST Shovel, 14 Ga., Double Rivet, Square Point, 48 in Polymer Handle
Catalog No. 502836741
Encompass
This shovel features a 14 gauge steel, square point blade with a double solid steel rivet head connection for durability. It is equipped with a 48 inch polymer handle that includes a fiberglass core for enhanced strength and comfort. The shovel is designed with a forward turned step for efficient digging and is suitable for various landscaping and construction tasks.
  • 14 gauge square point head
  • Double solid steel rivet head connection
  • Forward turned step
  • 48 inch polymer jacket with fiberglass core handle
  • Steel blade material
  • Overall length of 59.5 inches
Catalog No. 50-283-6741 Supplier MSC Supplier No. 52097631
This shovel features a 14 gauge steel, square point blade with a double solid steel rivet head connection for durability. It is equipped with a 48 inch polymer handle that includes a fiberglass core for enhanced strength and comfort. The shovel is designed with a forward turned step for efficient digging and is suitable for various landscaping and construction tasks.
  • 14 gauge square point head
  • Double solid steel rivet head connection
  • Forward turned step
  • 48 inch polymer jacket with fiberglass core handle
  • Steel blade material
  • Overall length of 59.5 inches