Learn More
Structure Probe Inc DBL SIDED ADH CNDTCV CARB DSCS
SDP
Catalog No. NC2080441
Encompass
Double Sided Adhesive Conductive Carbon Disc 9 mm Dia, 44 discs/sheet/10 sheets
Catalog No. NC2080441 Supplier Structure Probe Inc Supplier No. 05073BA QTY 19
Double Sided Adhesive Conductive Carbon Disc 9 mm Dia, 44 discs/sheet/10 sheets