Learn More
Electron Microscopy Sciences Secure-Seal™ Hybridization Double Sided Adhesive Sheet 0.24 mm thick, 7.25" x 10"
SDP
Catalog No. 5030450
Encompass_Preferred
SecureSeal adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides. Sealable access ports. Removes easily. All Hybridization Chambers have 1.5 mm diameter Ports and come with 120 Port Seals.
O.D.: 7.25" x 10"
Catalog No. 50-304-50 Supplier Electron Microscopy Sciences Supplier No. 7033302
SecureSeal adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides. Sealable access ports. Removes easily. All Hybridization Chambers have 1.5 mm diameter Ports and come with 120 Port Seals.
O.D.: 7.25" x 10"