Learn More
Structure Probe Inc Double Sided Adhesive Carbon Tape, 12 mm x 20 m, Plastic Core
SDP
Catalog No. NC9764204
Encompass
SPI Conductive Double Sided Carbon Adhesive Tape,12 mm wide x 20 meters,Enables the mounting of samples without contamination from either silver paint or carbon paint,Provide good conductivity
Catalog No. NC9764204 Supplier Structure Probe Inc Supplier No. 05082AB
SPI Conductive Double Sided Carbon Adhesive Tape,12 mm wide x 20 meters,Enables the mounting of samples without contamination from either silver paint or carbon paint,Provide good conductivity