Learn More
Electron Microscopy Sciences Conductive Double Sided Carbon Tape with Aluminum 5 mm 20 MT
SDP
Catalog No. 50365857
Encompass_Preferred
Can be used to mount particles 15-20 microns. Aluminum base.
Catalog No. 50-365-857 Supplier Electron Microscopy Sciences Supplier No. 7781705AL
Can be used to mount particles 15-20 microns. Aluminum base.