Learn More
Electron Microscopy Sciences Conductive Double Sided Carbon Tape with Aluminum 8 mm 20 MT
SDP
Catalog No. 50365968
Encompass_Preferred
Can be used to mount particles 15-20 microns. Aluminum base.
Catalog No. 50-365-968 Supplier Electron Microscopy Sciences Supplier No. 7781708AL
Can be used to mount particles 15-20 microns. Aluminum base.